Learn More
anti-Midkine, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP217289
Catalog No. NBP217289
Add to cart
Description
Description
Midkine Polyclonal specifically detects Midkine in Human samples. It is validated for Western Blot.
Specifications
Specifications
Midkine
1mg/mL
Western Blot 1:500-1:3000
P21741
Amphiregulin-associated protein, Midgestation and kidney protein, midkine, midkine (neurite growth-promoting factor 2), MK1, MKARAP, NEGF2FLJ27379, Neurite outgrowth-promoting factor 2, Neurite outgrowth-promoting protein
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human. Species reactivity based on sequence identity: Pig (94%), Rat (87%), Mouse (87%), Rabbit (94%), Cow (93%).
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Affinity Purified
MDK
Full length human Midkine Recombinant protein.
16 kDa
0.1 ml
Cell Cycle and Replication
Primary
4192
Human