Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIWIL1 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523438

Catalog No. PIPA523438


Add to cart

Description

Description

Mili, otherwise known as Piwi-like protein 2, is an Argonaute protein, a member of the ‘Piwi family.’ It is required for germ cell and stem cell development in invertebrates. Mili is essential for spermatogenesis in mouse and plays a central role during spermatogenesis by repressing transposable elements and preventing their mobilization, which is essential for the germline integrity. It is also essential for meiotic differentiation and self-renewal of spermatogonial stem cells.
Specifications

Specifications

PIWIL1
Polyclonal
Unconjugated
Piwil1
Liquid
PIWIL1
Synthetic peptide (44 GRGLVFRKPEDSSPPLQ 60) of Mili protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q9JMB7
CT80, HILI, PIWIL1L, mili, Miwi like, cancer/testis antigen 80, piwi-like protein 2, piwil2-like protein
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
57749
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit