Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL3 Rat anti-Mouse, APC, Clone: 39624, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA523647

Catalog No. PIMA523647


Add to cart

Description

Description

In Western blots, approximately 25% cross-reactivity with Recombinant mouse CCL4/MIP-1 beta is observed and no cross-reactivity with Recombinant human CCL1, 2, 3, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 21, 22, 23, 24, 25, 28, Recombinant mouse CCL1, 2, 6, 7, 9/10/MIP-1 gamma, 11, 12, 19, 21, 22, 25, 28, Recombinant rat CCL2 or CCL20 is observed.

Macrophage inflammatory protein-1a (MIP-1a) is an 8kDa member of the C-C chemokine subfamily. C-C chemokines are chemoattractants and activators for monocytes and T cells. MIP-1a has been identified as one of the major HIV-SFs produced by CD8+ T cells, along with MIP-1b and RANTES.
Specifications

Specifications

CCL3
Monoclonal
APC
Ccl3
Liquid
CCL3
E. coli-derived recombit mouse CCL3/MIP-1 alpha Ala24-Ala92
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
39624
Buffered saline solution with BSA and 0.09% sodium azide
P10855
C-C motif chemokine 3, G0/G1 switch regulatory protein 19-1, Macrophage inflammatory protein 1-alpha, PAT 464.1, SIS-beta, Small-inducible cytokine A3, Tonsillar lymphocyte LD78 alpha protein, MIP-1-alpha, LD78-alpha, CCL3, G0S19-1, MIP1A, SCYA3
Rat
IgG2A
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
20302
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit