Learn More
MIP-1b Mouse anti-Human, V450, Clone: D21-1351, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 561282
Catalog No. BDB561282
Add to cart
Description
Description
The D21-1351 monoclonal antibody specifically binds to the human CC chemokine, MIP-1β (macrophage inflammatory protein-1β). Human MIP-1β shares approximately 75% homology with mouse MIP-1β at the amino acid level. Expression of MIP-1β in human peripheral blood cells is induced by proinflammatory and mitogenic stimuli. MIP-1βis a chemoattractant for monocytes and lymphocytes. Human MIP-1β
The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450 nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Specifications
Specifications
MIP-1b
D21-1351
Macrophage inflammatory protein 1-beta; CCL4; C-C motif chemokine 4; LAG-1
Affinity Purified
Recombinant Human MIP-1β
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
V450
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Mouse
IgG1, κ
RUO
Primary
Human