Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIP-3 alpha Rat anti-Mouse, FITC, Clone: 114906, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA523548

Catalog No. PIMA523548

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Macrophage Inflammatory Protein-3 alpha (MIP-3 alpha), also called CCL20, is expressed in the liver, lungs, lymph nodes and peripheral blood lymphocytes. MIP-3 alpha is strongly up regulated by inflammatory signals, and down regulated by the anti-inflammatory cytokine IL-10. MIP3alpha signals through the G protein-coupled receptor, CCR6, and acts as a chemoattractant to lymphocytes and dendritic cells.
Specifications

Specifications

MIP-3 alpha
Monoclonal
FITC
Ccl20
Liquid
CCL20
E. coli-derived recombit mouse CCL20/MIP-3 alpha Ala28-Met97
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
114906
Buffered saline solution with BSA and 0.09% sodium azide
O89093
CKb4, LARC, MIP-3a, MIP3A, SCYA20, ST38, C-C motif chemokine 20, CC chemokine LARC, MIP-3-alpha, beta chemokine exodus-1, beta-chemokine exodus-1, exodus-1, liver and activation-regulated chemokine, macrophage inflammatory protein 3 alpha, small inducible cytokine subfamily A (Cys-Cys), member 20, small-inducible cytokine A20
Rat
IgG1
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
20297
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit