Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL2, Biotin, Polyclonal, Invitrogen™

Catalog No. PIPA132940
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA132940 50 μg
1 options

Catalog No. PIPA132940

Supplier: Thermo Scientific PA132940

Rabbit Polyclonal Antibody

PA1-32940 detects MIP2 from mouse samples.PA1-32940 has been successfully used in Western blot applications.The PA1-32940 immunogen is: Recombinant mouse MIP-2
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIP2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
Gene Cxcl2
Gene Accession No. P10889
Gene Alias CINC-2a, GROb, Gro2, MIP-2, MIP-2a, Mgsa-b, Mip2, Scyb, Scyb2
Gene Symbols CXCL2
Host Species Rabbit
Immunogen Recombinant mouse MIP-2
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20310
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.