Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitosin Mouse anti-Human, Unlabeled, Clone: 11, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610768

Catalog No. BDB610768


Add to cart

Description

Description

Mitosin, a nuclear proetin of 3113 amino acids, contains a pair of tandem repeats and seven leucine repeats. In vitro, Mitosin directly interacts with the retinoblastoma protein, Rb. Mitosin is expressed throughout the cell cycle, with levels being lowest during G1, and it localizes at the kinetochore during the mitotic phase. The subcellular redistribution of Mitosin to the kinetochore is linked to its phosphorylation. Its expression is linked to patients with autoimmune diseases characterized by abnormal cell proliferation. Ectopic expression of a truncated version of Mitosin blocks the progression of the cell cycle. This suggests that the protein has an important role during cell proliferation. Because of similarities in subcellular localization and expression, Mitosin and the autoantigen p300/CENP-F, which is also linked to a multitude of autoimmune disorders, are probably the same protein.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

Mitosin
11
Unconjugated
Affinity Purified
Human Mitosin aa. 209-381
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit