Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MNK1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513951

Catalog No. PIPA513951

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

MKNK1 encodes a predicted 424-amino acid protein containing a sequence typical of the catalytic domain of serine/threonine kinases and a putative N-terminal nuclear localization signal sequence.C-terminal region of MKNK1 was phosphorylated and activated in vivo and in vitro by ERK1 and p38 MAP kinases, but not by JNK/SAPK. The activated ERK is essential for the activation of MKNK1 in insect cells. MKNK1 was activated following stimulation of HeLa cells with a variety of extracellular stimuli. MNK1 bind tightly to the growth factor-regulated MAP kinases, Erk1 and Erk2, and also binds strongly to the stress-activated kinase, p38.
Specifications

Specifications

MNK1
Polyclonal
PBS with 0.09% sodium azide
Q9BUB5
MAP kinase-interacting serine/threonine-protein kinase 1; MAP kinase signal-integrating kinase 1; Mnk1; MKNK1; MNK1;
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8569
Western Blot
Unconjugated
MKNK1
Liquid
MKNK1
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human MKNK1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit