Learn More
MKP2 Mouse, Unlabeled, Clone: 48, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610850
Description
The mitogen activated protein (MAP) kinases mediate signal transduction pathways involved in cellular growth and differentiation. These MAP kinases, including ERK1 and ERK2, are activated by phosphorylation on Tyr and Thr by MEK (MAP and ERK Kinase). Regulation of the resulting physiological effects of MAP Kinase activation is affected in part by MAP Kinase phosphatases (MKPs). These phosphatases have dual specificity, dephosphorylating both the tyrosine and threonine residues on MAP kinases. MKP2, which is widely expressed in human tissues, specifically dephosphorylates activated ERKs and JNK. These phosphatases have overlapping substrate specificities for the three known families of MAP kinases - p38, ERK, and JNK. All of the known MKPs contain a highly conserved carboxyl-terminal catalytic domain flanked by two CH2 (CDC25 homology 2) domains.
Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
MKP2 | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Rat MKP2 aa. 13-127 | |
50 μg | |
Cell Biology | |
Frog, Human, Mouse, Rat | |
IgG1 |
Western Blot | |
48 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.