Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKRN3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525557

Catalog No. PIPA525557

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene contains a RING (C3HC4) zinc finger motif and several C3H zinc finger motifs.
Specifications

Specifications

MKRN3
Polyclonal
PBS with 0.09% sodium azide
Q13064
D15S9, RNF63, ZFP127, ZNF127, RING finger protein 63, probable E3 ubiquitin-protein ligase makorin-3, zinc finger protein 127
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7681
Western Blot
Unconjugated
MKRN3
Liquid
MKRN3
KLH conjugated synthetic peptide between 160-189 amino acids from the central region of human MKRN3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit