Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

myeloid leukemia factor 2, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human MLF2 protein.

Manufacturer:  Abnova Corporation H00008079B01

Catalog No. 89-006-535


Add to cart

Description

Description

Sequence: MFRFMRDVEPEDPMFLMDPFAIHRQHMSRMLSGGFGYSPFLSITDGNMPGTRPASRRMQQAGAVSPFGMLGMSGGFMDMFGMMNDMIGNMEHMTAGGNCQTFSSSTVISYSNTGDGAPKVYQETSEMRSAPGGIRETRRTVRDSDSGLEQMSIGHHIRDRAHILQRSRNHRTGDQEERQDYINLDESEAAAFDDEWRRETSRFRQQRPLEFRRLESSGAGGRRAEGPPRLAIQGPEDSPSRQSRRYDW
Specifications

Specifications

myeloid leukemia factor 2
Unconjugated
No additive
NM_005439
NTN4
Mouse
50 uL
Stem Cell Biology
Y
Polyclonal
Human
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human MLF2 protein.
MLF2
Serum
MLF2
MLF2 (NP_005430, 1 a.a. ~ 248 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
8079
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit