Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-MLH1 (Ab-2), Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ PC56100UG
Description
Specifically detects MLH1 Clone: in Human, Mouse samples, and it is validated for Immuno Blotting
The MLH1 gene is one of at least 4 genes encoding proteins involved in the repair of mismatch nucleotides following DNA replication or repair. Mutations in the MLH1 gene contribute to the development of sporadic colorectal carcinoma, while germline MLH1 mutations are responsible for approximately 35% of inherited, non-polyposis colorectal carcinoma (HNPCC). MLH1 encodes a protein of 756 amino acids.Specifications
MLH1 | |
Polyclonal | |
In 0.05M sodium phosphate buffer, 0.2% gelatin. | |
recombinant human MLH1 | |
RUO | |
Recognizes the ∽83kDa MLH1 protein. | |
2°C to 8°C | |
IgG |
Immunoblot | |
Unconjugated | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction