Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLK2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515107

Catalog No. PIPA515107

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

MLK2 is a member of the serine/threonine kinase family. This kinase has been shown to activate MAPK8/JNK and MKK4/SEK1, and this kinase itself can bephoshorylated, and thus activated by JNK kinases. This kinase functions preferentially on the JNK signaling pathway, and is reported to be involved in nerve growth factor (NGF) induced neuronal apoptosis.
Specifications

Specifications

MLK2
Polyclonal
Unconjugated
MAP3K10
Liquid
MAP3K10
KLH conjugated synthetic peptide between 628-659 amino acids from the C-terminal region of human MLK2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
1.9 mg/mL
PBS with 0.09% sodium azide
Q02779
Mixed lineage kinase 2; Mitogen-activated protein kinase kinase kinase 10; Protein kinase MST
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4294
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit