Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MLK4, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP141080

Catalog No. NBP141080


Add to cart

Description

Description

MLK4 Polyclonal antibody specifically detects MLK4 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications

Specifications

MLK4
1.0 mg/mL
Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative)
Affinity Purified
KIAA1804
The immunogen for this product maps to a region between residue 650 and 700 of human mixed lineage kinase 4 using the numbering given in entry NP_115811.2 (GeneID 84451).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
dJ862P8.3, EC 2.7.11, EC 2.7.11.25, mitogen-activated protein kinase kinase kinase MLK4, mixed lineage kinase 4, MLK4, RP5-862P8.2
Rabbit
IgG
100 ul
Protein Kinase
Primary
84451
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only