Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLK4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP141081
Description
MLK4 Polyclonal specifically detects MLK4 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
MLK4 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
dJ862P8.3, EC 2.7.11, EC 2.7.11.25, mitogen-activated protein kinase kinase kinase MLK4, mixed lineage kinase 4, MLK4, RP5-862P8.2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
Q5TCX8 | |
KIAA1804 | |
The immunogen for this product maps to a region between residue 986 and 1036 of human mixed lineage kinase 4 using the numbering given in entry NP_115811.2 (GeneID 84451). | |
0.1 mL | |
Protein Kinase | |
84451 | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction