Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MLLT1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP126652

Catalog No. NBP126652


Add to cart

Description

Description

MLLT1 Polyclonal antibody specifically detects MLLT1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications

Specifications

MLLT1
1.0 mg/mL
Immunoprecipitation 3-5 ug/mg lysate, Western Blot (Negative)
NP_005925.2
ENLENL/MLL fusion, LTG19MLLT1/MLL fusion, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 1, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 1, protein ENL, YEATS domain-containing protein 1, YEATS1myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocatedto, 1
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
MLLT1
The immunogen recognized by this antibody maps to a region between residue 150 and 200 of human myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 1 using the numbering given in entry NP_005925.2 (GeneID 4298).
Immunogen affinity purified
RUO
Primary
4298
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only