Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMAA Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526796

Catalog No. PIPA526796

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is involved in the translocation of cobalamin into the mitochondrion, where it is used in the final steps of adenosylcobalamin synthesis. Adenosylcobalamin is a coenzyme required for the activity of methylmalonyl-CoA mutase. Defects in this gene are a cause of methylmalonic aciduria.
Specifications

Specifications

MMAA
Polyclonal
PBS with 0.09% sodium azide
Q8IVH4, Q8C7H1
cblA, methylmalonic aciduria type A protein, mitochondrial
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
109136, 166785
Immunohistochemistry, Western Blot
Unconjugated
MMAA
Liquid
MMAA
KLH conjugated synthetic peptide between 56-84 amino acids from the N-terminal region of human MMAA
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit