Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMGT1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524667

Catalog No. PIPA524667

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

Mmgt1 encodes a protein that that mediates Mg(2+) transport.
Specifications

Specifications

MMGT1
Polyclonal
PBS with 0.09% sodium azide
Q8N4V1
TMEM32, transmembrane protein 32
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
93380
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
MMGT1
Liquid
MMGT1
KLH conjugated synthetic peptide between 103-131 amino acids from the C-terminal region of human MMGT1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit