Learn More
MilliporeSigma™ anti-MMP-1, hinge region, Polyclonal, ™
Supplier: MilliporeSigma™ AB6002/DEL
Description
All cells within tissues are surrounded by an extracellular matrix (ECM) giving the tissues shape and structure. The ECM is constantly being remodeled and constant communication is maintained between cells through this matrix. Secreted proteins, termed matrix metalloproteinases (MMPs), are involved in the modulation of cell matrix interactions. MMPs are Zn2+ binding endopeptidases that degrade various components of the ECM. MMPs are enzymes implicated in normal and pathologic tissue remodeling processes, wound healing, angiogenesis, and tumor invasion. These enzymes are very potent when active, and are associated with extracellular space inhibitors called TIMPs (tissue inhibitors of matrix metalloproteinases). MMP9, also known as gelatinase B, is a secreted enzyme which degrades the interstitial collagens, types I, II, and III and is produced by normal alveolar macrophages and granulocytes.
Host Species: Rabbit
Cross Reactivity: Human
Immunogen: KLH-conjugated synthetic peptide corresponding to the hinge region of human MMP-1.
Western Blotting
Specifications
MMP-1, hinge region | |
Unconjugated | |
KLH-conjugated synthetic peptide corresponding to the hinge region of human MMP-1. | |
100 μg | |
Human |
Polyclonal | |
Rabbit | |
Affinity Purified | |
Primary |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.