Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-13 Mouse anti-Human, Mouse, Rabbit, Clone: VIIIA2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB13424
Description
This Anti-MMP-13 Antibody, clone VIIIA2 is validated for use in immunohistochemistry for the detection of MMP-13.
Specifications
MMP-13 | |
Monoclonal | |
Unconjugated | |
NM_002427.2 | |
Mouse | |
100 μg | |
Human, Mouse, Rabbit | |
Purified |
Immunohistochemistry | |
VIIIA2 | |
Liquid in 10mM PBS, pH 7.4, with 0.2% BSA and 15mM sodium azide. | |
MMP13, MMP-13, CLG3, EC 3.4.24.- | |
Human recombinant collagenase-3 protein. | |
Primary | |
2-8°C in undiluted aliquots for up to 12 months. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction