Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MMP-13/Collagenase-3 Clone: 3H13L17, Invitrogen™

MMP-13/Collagenase-3 Monoclonal antibody 701287 from Thermo Scientific™ Pierce™ specifically detects MMP-13/Collagenase-3 in Human samples, and it is validated for WB, IF and ICC

Manufacturer:  Invitrogen 701287

Encompass

Catalog No. 70-128-7


Add to cart

Description

Description

Matrix metallopeptidase 13 belongs to the matrix metalloproteinase peptidase M10A subfamily. The inactive polypeptide undergoes serine protease cleavage, resulting in an active form containing catalytic and hemopexin-like domains. MMP-13 is involved in the breakdown of the extracellular matrix during tissue remodeling, embryonic development, and reproduction. MMP-13 cleaves type II collagen more efficiently than types I and III.

701287 detects MMP-13/Collagenase-3 in Human samples. 701287 has been successfully used in WB, IF and ICC procedures. The immunogen for 701287 is recombinant protein corresponding to amino acids 221-471 of human MMP-13.

Applications:

Immunocytochemistry, Immunofluorescence, Western Blotting

Specifications

Specifications

MMP-13/Collagenase-3
Unconjugated
P45452
Rabbit
IgG
Store at 2 to 8°C for up to 1 month. For long term storage, store at -20°C.
Monoclonal
Human
3H13L17
100μg of Protein A antibody in PBS with 0.09% sodium azide;
Purified
Recombinant protein corresponding to amino acids 221-471 of human MMP-13
Protein A
Primary
4322
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit