Learn More
MMP-2 Mouse anti-Chicken, Human, Clone: 7F3.2, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma MAB13435
Catalog No. MAB13435MI
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-MMP-2 Antibody, clone 7F3.2 is an antibody against MMP-2 for use in ELISA and western blotting.
Specifications
Specifications
MMP-2
Monoclonal
Unconjugated
NM_004530.2
Mouse
100 μg
Chicken, Human
Purified
ELISA, Western Blot
7F3.2
Liquid at 1mg/mL containing sodium azide as a preservative.
MMP2, TBE-1, CLG4A, CLG4, MMP-II, MONA, MMP-2, EC 3.4.24.24
Synthetic peptide based on the human sequence.
Primary
2-8°C undiluted for 6 months.
IgG1