Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP3 Mouse anti-Human, Clone: SL-1 IID4, Invitrogen
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MS810PABX
Description
MS810PABX targets MMP-3 in AP, IF, and WB applications and shows reactivity with Human samples. The MS810PABX immunogen is aPMA (4-aminophenylmercuric acetate) activated human MMP-3. MS810PABX detects MMP-3 which has a predicted molecular weight of approximately 43 kDa.
MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form, and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions, and has metallo-endopeptidase activity. The active enzyme degrades collagen, gelatin, fibronectin, and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.Specifications
MMP3 | |
Monoclonal | |
Unconjugated | |
MMP3 | |
matrix metalloproteinase 3 (stromelysin 1, progelatinase); matrix metalloproteinase-3; proteoglycanase; stromelysin-1; transin-1 | |
Mouse | |
Protein A | |
RUO | |
4314 | |
-20°C | |
Liquid |
Affinity Purification, Immunofluorescence, Immunohistochemistry, Western Blot | |
SL-1 IID4 | |
PBS with no preservative; pH 7.4 | |
P08254 | |
MMP3 | |
APMA (4-aminophenylmercuric acetate) activated human MMP-3 | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction