Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP14, Polyclonal, Invitrogen™

Catalog No. PIPA127560
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127560 50 μg
1 options

Catalog No. PIPA127560

Supplier: Thermo Scientific PA127560

Chicken Polyclonal Antibody

Seems to specifically activate progelatinase A. May thus trigger invasion by tumor cells by activating progelatinase A on the tumor cell surface. Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix i

PA1-27560 detects MMP14 from human and rat samples.PA1-27560 may be successfully used in Western blot applications.The immunogen is a synthetic peptide corresponding to human Matrix metalloproteinase-14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP14
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene MMP14
Gene Accession No. P50281, Q10739
Gene Alias MMP-X1, MT1-MMP, MTMMP1
Gene Symbols MMP14
Host Species Chicken
Immunogen Synthetic peptide corresponding to human Matrix metalloproteinase-14.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4323
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.