Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP14 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138193

Catalog No. 01-673-900


Add to cart

Description

Description

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is placenta or breast carcinoma.

MMPs are a group of enzymes involved in matrix degradation. Of the sixteen members of the matrix metalloproteinase family, ten exist in soluble form whereas MT-MMPs exist as integral membrane proteins. MT1-MMP, MT2-MMP, MT3-MMP also known as MMP14, MMP15, MMP16 respectively, contain a C-terminal transmembrane domain anchoring them to the cell surface. They have an 8 amino acid insert in their catalytic domain. MT1-MMP cleaves progelatinase A (MMP-2) and progelatinase B to their active forms. MT1-MMP gets activated through the membrane plasmin cascade. It binds to TIMP-2 first and then to MMP-2 forming a trimer. It cleaves the pro-MMP-2 to its active form.
Specifications

Specifications

MMP14
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.6
P50281
1, MMP-14, MMP-X1, MT-MMP, MT-MMP 1, MT1-MMP, MT1MMP, MTMMP1, matrix metalloproteinase 14 membrane-inserted, matrix metalloproteinase-14, membrane type 1 metalloprotease, membrane-type matrix metalloproteinase 1, membrane-type-1 matrix metalloproteinase
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4323
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
MMP14
Liquid
MMP14
Synthetic peptide derived from internal region of human MMP-14 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit