Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP23B, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA125237

Catalog No. PIPA125237

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

PA1-25237 detects MMP-22 from chicken samples.PA1-25237 has been successfully used in Western blot procedures.The PA1-25237 immunogen is a synthetic peptide corresponding to the C-terminal region of chicken MMP-22.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MMP22
Polyclonal
Unconjugated
MMP23A
Rabbit
Antigen affinity chromatography
RUO
419407
Store at -20°C, Avoid Freeze/Thaw Cycles
Purified
Western Blot
1mg/mL
MMP23B
MMP23B
Synthetic peptide corresponding to the C-terminal region of chicken MMP-22.
25 μg
Primary
Chicken
Antibody
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit