Learn More
modulator of apoptosis 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MOAP1 protein.
Supplier: Abnova Corporation H00064112B01
Description
The protein encoded by this gene was identified by its interaction with apoptosis regulator BAX protein. This protein contains a Bcl-2 homology 3 (BH3)-like motif, which is required for the association with BAX. When overexpressed, this gene has been shown to mediate caspase-dependent apoptosis. [provided by RefSeq
Sequence: MTLRLLEDWCRGMDMNPRKALLIAGISQSCSVAEIEEALQAGLAPLGEYRLLGRMFRRDENRKVALVGLTAETSHALVPKEIPGKGGIWRVIFKPPDPDNTFLSRLNEFLAGEGMTVGELSRALGHENGSLDPEQGMIPEMWAPMLAQALEALQPALQCLKYKKLRVFSGRESPEPGEEEFGRWMFHTTQMIKAWQVPDVEKRRRLLESLRGPALDVIRVLKINNPLITVDECLQALEEVFGVTDNPRELQVKYLTTYQKDEEKLSAYVLRLEPLLQKLVQRGAIERDAVNQARLDQVIAGAVHKTIRRELNLPEDGPAPGFLQLLVLIKDYEAAEEEEALLQAILEGNFTSpecifications
modulator of apoptosis 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human MOAP1 protein. | |
MOAP1 | |
MAP-1/PNMA4 | |
Mouse | |
50 μL | |
Apoptosis | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
NM_022151 | |
MOAP1 | |
MOAP1 (NP_071434, 1 a.a. ∼ 351 a.a) full-length human protein. | |
RUO | |
Yes | |
64112 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.