Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOCS1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523759

Catalog No. PIPA523759

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

MONDOA forms heterodimers with MLX (MIM 602976) that can bind to and activate transcription from CACGTG E boxes.
Specifications

Specifications

MOCS1
Polyclonal
PBS with 0.09% sodium azide
Q9NZB8, Q5RKZ7
MIR, MONDOA, bHLHe36, MLX-interacting protein, Mlx interactor, MondoA, class E basic helix-loop-helix protein 36, transcriptional activator MondoA
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4337, 56738
Immunohistochemistry, Western Blot
Unconjugated
MOCS1
Liquid
MOCS1
KLH conjugated synthetic peptide between 462-490 amino acids from the C-terminal region of human MOCS1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit