Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-MONKEY IGG AB FITC  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52300422
Affinity purified polyclonal antibody to Monkey IgG, both heavy and light chains (whole IgG) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-671-87