Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monkey IgG Isotype Control, Biotin, Novus Biologicals™
SDP

Monkey Antibody

Supplier:  Novus Biologicals NBP197172

Catalog No. NBP197172


Add to Cart

Description

Description

IgG specifically detects IgG in Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications

Specifications

IgG
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
IGHG1
Multi-step
RUO
3500
Reconstitute with 500 ul deionized water (or equivalent).
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
IgG
Biotin
Immunoglobulin G, ImmunoglobulinG
Monkey
0.5 mg
Isotype Control (Primary)
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Monkey IgG and anti-Monkey Serum.
Monkey
Lyophilized
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only