Learn More
anti-Monkey IgG, Fluorescein, Isotype Control, Novus Biologicals™
Monkey Isotype Control Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197150
Catalog No. NBP197150
Add to cart
Description
Description
IgG Isotype Control antibody specifically detects IgG in Monkey samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
IgG
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Sodium Azide
immunoglobulin heavy constant gamma 1 (G1m marker)
Monkey
Ion exchange chromatography
RUO
Isotype Control
3500
Reconstitute with 0.5 ml deionized water (or equivalent) to desired concentration.
Fluorescein
Lyophilized
IGHG1
IgG
0.5 ml
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Fluorescein, anti-Monkey IgG and anti-Monkey Serum.
Monkey