Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-MONKEY IGG (GAMMA) AB HRP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200333
Affinity purified polyclonal antibody to Monkey IgG heavy chains (gamma) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-671-98