Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-MONKEY IGM (U) AB  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52100217
Affinity purified polyclonal antibody to Monkey IgM heavy chains (mu) made in Goat. Product is in liquid form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-670-31