Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Monocyte/Granulocyte Mouse anti-Porcine, R-PE, Clone: 74-22-15, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 452509
Description
SouthernBiotech anti-porcine primary antibodies are available in a variety of conjugated formats including Alexa Fluor.
Specifications
Monocyte/Granulocyte | |
Monoclonal | |
0.1mg/mL | |
Supplied in PBS/NaN3 and a stabilizing agent | |
Fresh dd miniature swine thymocytes | |
RUO | |
Porcine | |
IgG1 |
Flow Cytometry | |
74-22-15 | |
R-PE | |
Mouse | |
0.1 mg | |
Primary | |
Store at 2-8°C; Protect from light; Do not freeze |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title