Learn More
Monomethyl-Histone H3 (Lys27) Rabbit anti-Human, AlexaFluor™488, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma FCABS304A4
Catalog No. FCABS304A4
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect monomethyl-Histone H3 (Lys27) using this Milli-Mark™ Anti-monomethyl-Histone H3 (Lys27)-Alexa Fluor 488 Antibody validated for use in flow cytometry.
Specifications
Specifications
monomethyl-Histone H3 (Lys27)
Alexa Fluor 488
Purified
Rabbit
IgG
100 Tests
Primary
NP_003484
Flow Cytometry
Purified rabbit polyclonal IgG conjugated to Alexa Fluor 488 in PBS with 0.1% sodium azide and 15mg/mL BSA.
HIST3H3; H3FT; H3/t; H3t; MGC126888; H3/g; H3.4; H3T; MGC126886
KLH-conjugated, 2X-branched synthetic peptide corresponding to monomethyl-lysine at residue 27 of human Histone H3.
Protein A Purified
2-8°C protected from light for up to 6 months
Polyclonal
Human