Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monomethyl-Histone H3 (Lys27) Rabbit anti-Human, Vertebrates, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$119.20 - $499.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen KLH-conjugated, synthetic 2X-branched peptide containing the sequence ...ARmeKSA... in which meK corresponds to monomethyl-lysine at residue 27 of human histone H3.
Antigen monomethyl-Histone H3 (Lys27)
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
07-448-SMI
View Documents
MilliporeSigma
07448S
20μg
Each for $119.20
Add to cart
 
07-448-MI
View Documents
MilliporeSigma
07448
200μg
Each for $499.00
Add to cart
 
Description

Description

Anti-monomethyl-Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody to detect Histone H3 monomethylated at lysine 27. This purified Ab, also known as Anti-H3K27me1 is specificity verified by dot blot (DB), published in peer reviewed journals and validated in immunocytochemistry, western blot, Chimmunoprecipitation, ChIP-seq.
Specifications

Specifications

Rabbit
KLH-conjugated, synthetic 2X-branched peptide containing the sequence ...ARmeKSA... in which meK corresponds to monomethyl-lysine at residue 27 of human histone H3.
Polyclonal
Primary
NP_003484
IgG
monomethyl-Histone H3 (Lys27)
Purified
ChIP-sequencing, Dot Blot, Immunocytochemistry, Western Blot
Purified rabbit IgG in buffer containing 0.01M phosphate buffer, pH 7.6, 0.125M NaCl, 0.05% sodium azide and 30% glycerol.
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit