Learn More
anti-monomethyl-Histone H3 (Lys27), Polyclonal, EMD Millipore™ Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Millipore Sigma 07448
 View more versions of this product
Catalog No. 07-448-MI
Add to cart
Description
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
monomethyl-Histone H3 (Lys27)
Unconjugated
Purified
Rabbit
IgG
200μg
Primary
NP_003484
ChIP-sequencing, Dot Blot, Immunocytochemistry, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
KLH-conjugated, synthetic 2X-branched peptide containing the sequence ...ARmeKSA... in which meK corresponds to monomethyl-lysine at residue 27 of human histone H3.
Protein A purfied
2-8°C for 12 months
Polyclonal
Human, Vertebrate