Learn More
Monomethyl-Histone H3 (Lys9) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061416
Catalog No. 06-141-6MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-monomethyl-Histone H3 (Lys9) Antibody is a rabbit polyclonal antibody for detection of monomethyl-Histone H3 (Lys9) also known as H3K9me1, Histone H3 (mono methyl K9), H3me3K9 and has been validated in western blot, immunocytochemistry, PIA.
Specifications
Specifications
monomethyl-Histone H3 (Lys9)
Unconjugated
Affinity Purified
Rabbit
Affinity Purfied
2-8°C for 12 months
Polyclonal
Human
Immunocytochemistry, Inhibition Assays, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
HIST3H3; H3FT; H3/t; H3t; H3/g; H3.4; H3T
KLH-conjugated linear peptide corresponding to human Histone H3 methylated at and around Lys9.
100μg
Primary
NP_003484.1