Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Monomethyl-Histone H3 (Lys9) Mouse anti-Human, Clone: CMA306, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051248S
Description
Use Anti-monomethyl Histone H3 (Lys9) Antibody, clone CMA306 (mouse monoclonal antibody) validated in western blot, PIA to detect monomethyl Histone H3 (Lys9) also known as H3K9me1, Histone H3 (mono methyl K9) and H3-K9-HMTase 1Specifications
monomethyl Histone H3 (Lys9) | |
Monoclonal | |
Unconjugated | |
NP_003484 | |
Mouse | |
Protein G purfied | |
Primary | |
Purified |
Inhibition Assays, Western Blot | |
CMA306 | |
Purified mouse monoclonal IgG2a in PBS with 0.05% sodium azide. | |
EC 2.1.1.43, KMT1A , MG44, OTTHUMP00000024298, SUV39H | |
KLH conjugated linear peptide corresponding to monomethylated Lys9 at the N-terminal region of Histone H3. | |
10 μg | |
Human | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only