Learn More
MilliporeSigma™ anti-MOP3 (BMAL1); Polyclonal; ™ (Chemicon™)
Supplier: MilliporeSigma™ AB4140/DEL
Description
Immunocytochemistry, Immunohistochemistry, Western Blotting
Specifications
MOP3 (BMAL1) | |
Unconjugated | |
Synthetic peptide corresponding to amino acids 257-272 of human MOP3. (SVKVEDKDFPSTCSKK). | |
100 μg | |
Neuroscience | |
Human, Mouse, Rat |
Polyclonal | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary |