Learn More
BMAL1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA132219
Catalog No. PIPA132219
Add to cart
Description
Description
PA1-32219 detects MOP3 from human samples. PA1-32219 has been successfully used in Western blot applications. The PA1-32219 immunogen is: Bacterially expressed human MOP3
The protein encoded by this gene is a basic helix-loop-helix protein that forms a heterodimer with CLOCK. This complex binds an E-box upstream of the PER1 gene, activating this gene and possibly other circadian rhythym-associated genes. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
Specifications
BMAL1
Polyclonal
whole serum with 0.02% sodium azide
O00327
BMAL1, BMAL1c, JAP3, MGC47515, MOP3, PASD3, TIC, bHLHe5
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Western Blot
Unconjugated
ARNTL
Liquid
ARNTL
Bacterially expressed human MOP3
400 μL
4° C, do not freeze
Primary
406