Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MORF4, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127845

Catalog No. PIPA127845

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Cellular senescence. the terminal nondividing state that normal cells enter following completion of their proliferative potential. is the dominant phenotype in hybrids of normal and immortal cells. Fusions of immortal human cell lines with each other have

PA1-27845 detects MORF4 from human and mouse samples.PA1-27845 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Mortality factor 4.
Specifications

Specifications

MORF4
Polyclonal
PBS with no preservative
Q9UBU8, P60762
CSR, CSRB, SEN, SEN1
Chicken
IgY
50μg
Store at -20°C
Primary
10933, 10934
Western Blot
Unconjugated
MORF4L1
Purified
MORF4, MORF4L1
Synthetic peptide corresponding to human Mortality factor 4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit