Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOSC1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526758

Catalog No. PIPA526758

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

MOSC1
Polyclonal
PBS with 0.09% sodium azide
Q5VT66
MOSC1, MOCO sulphurase C-terminal domain containing 1, MOSC domain-containing protein 1, mitochondrial, moco sulfurase C-terminal domain-containing protein 1, molybdenum cofactor sulfurase C-terminal domain-containing protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64757
Immunohistochemistry, Western Blot
Unconjugated
MARC1
Liquid
MARC1
KLH conjugated synthetic peptide between 175-204 amino acids from the central region of human MOSC1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit