Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Motilin Rabbit anti-Feline, Human, Porcine, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA136122

Catalog No. 01-672-283


Add to cart

Description

Description

PA1-36122 does not react with rat samples. A recommended positive control for IHC is pig duodenum. Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide.

This gene encodes a small peptide hormone that is secreted by cells of the small intestine to regulate gastrointestinal contractions and motility. Proteolytic processing of the secreted protein produces the mature peptide and a byproduct referred to as motilin-associated peptide (MAP). Two transcript variants encoding different preproprotein isoforms but the same mature peptide have been found for this gene.
Specifications

Specifications

Motilin
Polyclonal
whole serum with 0.1% sodium azide
Q9XSE2, P12872, P01307
MGC138519
Rabbit
IgG
RUO
Antibody
Polyclonal
Feline, Human, Porcine
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
MLN
Lyophilized
MLN
Proprietary immunogen.
50 μL
4° C
Primary
397466, 4295, 493834
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit