Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
anti-Mouse 14-3-3 eta, Alexa Fluor 405, Clone: 3G12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192691AF405
Catalog No. NBP192691W
Description
Description
14-3-3 eta Monoclonal antibody specifically detects 14-3-3 eta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
14-3-3 eta
3G12
Western Blot, Immunocytochemistry/Immunofluorescence
Q04917
14-3-3 protein eta, Protein AS1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, etapolypeptide, YWHA114-3-3 eta
Mouse
IgG1
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
Purified
YWHAH
Full length recombinant human 14-3-3 eta [UniProt# Q04917].
28 kDa
0.1 ml
Apoptosis, Cancer, Neuroscience
Primary
7533