Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-14-3-3 eta, Clone: 3G12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192691SS

 View more versions of this product

Catalog No. NBP192691SS


Add to cart

Description

Description

14-3-3 eta Monoclonal antibody specifically detects 14-3-3 eta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

14-3-3 eta
3G12
Unconjugated
50% PBS, 50% Glycerol with 5mM Sodium Azide
Purified
YWHAH
Full length recombinant human 14-3-3 eta [UniProt# Q04917].
28 kDa
0.025 ml
Apoptosis, Cancer, Neuroscience
Primary
7533
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
1 mg/mL
Western Blot 1:5000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000
Q04917
14-3-3 protein eta, Protein AS1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, etapolypeptide, YWHA114-3-3 eta
Mouse
IgG1
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only