Learn More
14-3-3 eta Antibody (3G12), DyLight 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP192691UV
Catalog No. NBP192691UV
Add to Cart
Description
Description
14-3-3 eta Monoclonal specifically detects 14-3-3 eta in Human, Mouse, Rat, Bovine, Canine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
Specifications
14-3-3 eta
Monoclonal
DyLight 350
Q04917
YWHAH
Full length recombinant human 14-3-3 eta expressed in and purified from E.coli [UniProt# Q04917].
Protein G purified
RUO
Primary
Human, Mouse, Rat, Bovine, Canine, Equine
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay
3G12
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated
14-3-3 protein eta, Protein AS1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, etapolypeptide, YWHA114-3-3 eta
Mouse
28 kDa
0.1 mL
Apoptosis, Cancer, Neuroscience
7533
Store at 4C in the dark.
IgG1
Product Suggestions
Product Suggestions