Learn More
anti-14-3-3 zeta/delta, Clone: 1314CT423.108.153.173.140, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232739
Catalog No. NBP232739
Add to cart
Description
Description
14-3-3 zeta/delta Monoclonal antibody specifically detects 14-3-3 zeta/delta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
14-3-3 zeta/delta
1314CT423.108.153.173.140
Unconjugated
PBS with 0.09% Sodium Azide
Purified
YWHAZ
This YWHAZ antibody is generated from a mouse immunized with a recombinant protein from human YWHAZ.
27.745 kDa
400 ul
Cell Cycle and Replication, Phospho Specific
Primary
7534
Western Blot, Immunocytochemistry, Immunofluorescence
0.44 mg/mL
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500
P63104, P63104
KCIP-1MGC126532,14-3-3-zeta, MGC111427, MGC138156, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, deltapolypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zetapolypeptide, zeta polypeptide
Mouse
IgG2b Kappa
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat