Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse 6-Histidine Epitope Tag, HRP, Clone: 33D10.D2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142783

Catalog No. NBP142783


Add to cart

Description

Description

His Tag Monoclonal antibody specifically detects His Tag in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

His Tag
33D10.D2
HRP
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Gentamicin Sulfate
6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine
His Tag antibody was produced in mice by repeated immunizations with 6X His epitope tag peptide H-H-H-H-H-H conjugated to KLH using maleimide.
Protein A purified
RUO
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with 100 ul deionized water.
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
LYOPH
Western Blot 1:1000-1:5000, ELISA 1:10000, Immunohistochemistry 1:500-1:2500, Immunohistochemistry-Paraffin 1:10-1:500
Affinity Purified
Mouse
IgG1 Kappa
0.1 mg
Epitope Tags, His Epitope Tags
Primary
6X His tag specific.
Non-species specific
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only