Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse A20/TNFAIP3, PE, Clone: 59A426, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP177533PE

Catalog No. NBP177533PE


Add to cart

Description

Description

A20/TNFAIP3 Monoclonal antibody specifically detects A20/TNFAIP3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

A20/TNFAIP3
59A426
Flow Cytometry
P21580
A20TNFA1P2, EC 3.4.19.12, EC 6.3.2.-, MGC104522, MGC138687, OTU domain-containing protein 7C, OTUD7CMGC138688, Putative DNA-binding protein A20, TNF alpha-induced protein 3, tumor necrosis factor alpha-induced protein 3, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha-induced protein 3, Zinc finger protein A20
Mouse
IgG1 Kappa
0.1 ml
Cancer, Signal Transduction
Primary
7128
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
Purified
TNFAIP3
Full length recombinant human A20. The epitope has been mapped to the C-terminal portion of A20 (amino acids 440-790)
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only